Key Documents
22327
Stainless Steel Screening
pkg of 1 ea
About This Item
Polecane produkty
opakowanie
pkg of 1 ea
Opis ogólny
An alternative to the use of wool plugs to hold packing material inside columns. Includes a single 2 in. x 2 in. (5 cm x 5 cm) sheet of 10 μm pore size material.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)