Przejdź do
22050
SWAGELOK® Nut, Front and Back Ferrule Set
SWAGELOK® 100-S, stainless steel, 1/16 in. SWAGELOK
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
137,00 zł
Informacje o tej pozycji
eCl@ss:
32035302
UNSPSC Code:
41115717
NACRES:
SB.54
137,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Pozwól nam pomóc
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
SWAGELOK® 100-S
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/16 in. SWAGELOK
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. Then slide on the nut with the open side facing the end of the tubing. Next, slide on the back ferrule with the wider part facing the nut. In the next step, slide on the front ferrule with the small end of the cone facing the end of the tubing. Then insert the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
220142202422029
description
SWAGELOK® 100-S, stainless steel, 1/16 in. SWAGELOK
description
SWAGELOK® 200-S, brass, 1/8 in. SWAGELOK
description
SWAGELOK® 100-S, brass, 1/16 in. SWAGELOK
description
SWAGELOK® 400-S, stainless steel, 1/4 in. SWAGELOK
fitting
1/16 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/16 in. SWAGELOK
fitting
1/4 in. SWAGELOK
packaging
pkg of 1 ea
packaging
pkg of 10 ea
packaging
pkg of 10 ea
packaging
pkg of 5 ea
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
-
technique(s)
gas chromatography (GC): suitable using :Suitable
manufacturer/tradename
SWAGELOK® 100-S
manufacturer/tradename
SWAGELOK® 200-S
manufacturer/tradename
SWAGELOK® 100-S
manufacturer/tradename
SWAGELOK® 400-S
material
stainless steel
material
brass
material
brass
material
stainless steel
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)