Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 pkg | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 331,00 zł |
Informacje o tej pozycji
UNSPSC Code:
41115717
NACRES:
SB.54
eCl@ss:
27150601
331,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
brass
packaging
pkg of 10 ea
manufacturer/tradename
SWAGELOK® 100-S
fitting
1/16 in. SWAGELOK
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. While assembling a compression tube fitting, the metal tubing should be inspected properly to ensure that the surface is smooth. Then slide on the nut with the open side facing the end of the tubing. Next, slide on the back ferrule with the wider part facing the nut. In the next step, slide on the front ferrule with the small end of the cone facing the end of the tubing. Then insert the tubing into the fitting and tighten up the nut/ferrule assembly onto the fitting.
Application
Swagelok® nut, front and back ferrule set maybe used for sampler construction from a single capillary gas chromatographic column, for collection of semi-volatile organic compounds through a diffusion denuder.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description SWAGELOK® 100-S, brass, 1/16 in. SWAGELOK | description SWAGELOK® 200-S, brass, 1/8 in. SWAGELOK | description SWAGELOK® 400-S, brass, 1/4 in. SWAGELOK | description SWAGELOK® 100-S, stainless steel, 1/16 in. SWAGELOK |
| fitting 1/16 in. SWAGELOK | fitting 1/8 in. SWAGELOK | fitting 1/4 in. SWAGELOK | fitting 1/16 in. SWAGELOK |
| packaging pkg of 10 ea | packaging pkg of 10 ea | packaging pkg of 10 ea | packaging pkg of 1 ea |
| material brass | material brass | material brass | material stainless steel |
| manufacturer/tradename SWAGELOK® 100-S | manufacturer/tradename SWAGELOK® 200-S | manufacturer/tradename SWAGELOK® 400-S | manufacturer/tradename SWAGELOK® 100-S |
Still not finding the right product?
Explore all of our products under Swagelok® Nut, Front and Back Ferrule Set
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?