Przejdź do
22038
SWAGELOK® Front Ferrule
SWAGELOK® 203-1, stainless steel, 1/8 in. SWAGELOK
Synonim(y):
Swagelok® 203-1,SS FRT FERR 1/8" PK5
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
5 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
114,00 zł
Informacje o tej pozycji
UNSPSC Code:
41115717
NACRES:
SB.54
114,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Pozwól nam pomóc
material
stainless steel
packaging
pkg of 5 ea
manufacturer/tradename
SWAGELOK® 203-1
technique(s)
gas chromatography (GC): suitable using :Suitable
fitting
1/8 in. SWAGELOK
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Brass ferrules are used with copper tubing whereas stainless-steel ferrules are used with stainless-steel tubing. Furthermore, the stainless steel ferrules have great mechanical strength, but they cannot be removed once they are on the column.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
220272203922012
description
SWAGELOK® 203-1, stainless steel, 1/8 in. SWAGELOK
description
SWAGELOK® 403-1, stainless steel, 1/4 in. SWAGELOK
description
SWAGELOK® 204-1, stainless steel, 1/8 in. SWAGELOK
description
SWAGELOK® 203-1, brass, 1/8 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/4 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/8 in. SWAGELOK
packaging
pkg of 5 ea
packaging
pkg of 5 ea
packaging
pkg of 5 ea
packaging
pkg of 20 ea
material
stainless steel
material
stainless steel
material
stainless steel
material
brass
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
technique(s)
gas chromatography (GC): suitable using :Suitable
manufacturer/tradename
SWAGELOK® 203-1
manufacturer/tradename
SWAGELOK® 403-1
manufacturer/tradename
SWAGELOK® 204-1
manufacturer/tradename
SWAGELOK® 203-1
Still not finding the right product?
Explore all of our products under Swagelok® Front Ferrule
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)