Merck
Z765597
Tube rack for Benchmixer™ XL multi-tube vortexer
Holds 15 x 50 mL tubes
Informacje o tej pozycji
NACRES:
NB.12
UNSPSC Code:
41122804
Manufacturer/tradename:
Benchmark Scientific BV1010-500
Feature:
Holds 15 x 50 mL tubes
Packaging:
pkg of 1 ea
Pomoc techniczna
Pozwól nam pomóc
feature
Holds 15 x 50 mL tubes
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-500
General description
Benchmixer XL vortexer head adapters to hold multiple tubes.
This rack enables the use of 15 x 50 mL tubes in the BenchMixer™ XL heavy-duty hands-free mixing vortex mixer. It is ideal for the QuEChERS pesticide analysis method.
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)