Przejdź do zawartości
Z757381
Sartorius Proline® Plus mechanical pippettor
single channel, volume 20-200 μL
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
20-200 μL
Packaging:
pack of 1 ea
Feature:
electronic: no, single-channel
Manufacturer/tradename:
Sartorius 728060
Pomoc techniczna
Pozwól nam pomóc
feature
electronic: no, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728060
volume
20-200 μL
General description
The Sartorius Proline Plus pipettes of the Biohitfamily are high quality with a light pipetting force. With robust design, the Proline Plus is suitable for variable every day laboratory applications and covers the full range of 0.1μL to 10 mL.
  • Reduced pipetting forces prevent RSI and improve the results
  • Accurate and precise
  • Contamination free pipetting with Safe-Cone Filters
  • Robust design
  • Easiest pipette to maintain
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)