Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2050,00 zł |
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
1-10 mL
Packaging:
pack of 1 ea
Feature:
electronic: no, single-channel
Manufacturer/tradename:
Sartorius 725090
2050,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócfeature
electronic: no, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 725090
volume
1-10 mL
General description
Sartorius′s new mechanical pipettor family - the mLINE - offers a unique combination of novel features for mechanical pipetting. Excellent performance, full autoclavability, smooth ergonomics and perfect safety. The mLINE pipettors cover the full volume range of 0.1 to 5000.
- Extremely light pipetting and tip ejection
- Unique safety features
- Stepless volume adjustment and patented volume locking system
- Thermal insulation of internal components to improve accuracy
- Full autoclavability without disassembling
- Easy cleaning, maintenance and calibration
- ID tag and color-coded caps
- Safe-Cone Filters and unique ejection
- Opti-load tip mounting mechanism for even tip sealing and light tip ejection in multichannel pipettors
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description single channel, volume 1-10 mL | description single channel, volume 100-1000 μL | description Single channel, volume 10-100 μL | description Single channel, volume 2-20 μL |
| manufacturer/tradename Sartorius 725090 | manufacturer/tradename Sartorius 725070 | manufacturer/tradename Sartorius 725050 | manufacturer/tradename Sartorius 725030 |
| feature single-channel, electronic: no | feature electronic: no, single-channel | feature single-channel, electronic: no | feature electronic: no, single-channel |
| volume 1-10 mL | volume 100-1000 μL | volume 100 μL | volume 20 μL |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Sartorius mLINE® mechanical pipettor
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?