Przejdź do
Z743581
Titer Tops® sealing film for microplates
pack of 50
Synonim(y):
microplate sealing film, multiwell plate sealing film, plate seal, Diversified Biotech T-TOPS-50, laboratory sealing film
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
50 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
233,00 zł
Informacje o tej pozycji
NACRES:
NB.17
UNSPSC Code:
41103032
Manufacturer/tradename:
Diversified Biotech T-TOPS-50
Sterility:
non-sterile
Packaging:
pack of 50
233,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
sterility
non-sterile
feature
prevents well-to-well contamination, sample spillage and evaporation
packaging
pack of 50
manufacturer/tradename
Diversified Biotech T-TOPS-50
Features and Benefits
  • Durable and thick (4.4 mil) yet easily pierced with a pipette tip
  • Prevents well-to-well contamination, sample spillage and evaporation
  • Polyethylene film that provides excellent adhesion to plastic microplates
  • Non-cytotoxic, sterile, solvent resistant and waterproof
  • Stays solidly in place even when exposed to 100% humidity and extreme temperatures ranging from -40 °C to +100 °C (Not recommended for use in thermal cyclers)
  • Chemically resistant and thermally stable
  • Easily marked with any writing instrumentTwo scored end taps for easy positioning
  • UV transparent down to 250 nm
Pre-cut, pierceable polyethylene microplate film for preventing cross-well contamination. End tabs make film positioning and removal hassle-free.
Legal Information
Titer-Tops is a registered trademark of Diversified Biotech
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z744132Z688630Z743564
description
pack of 50
description
polyester, W × L 3.125 in. × 5.750 in.
description
pk of 100 ×
description
clear, L × W 12.5 in. × 32.9 in.
manufacturer/tradename
Diversified Biotech T-TOPS-50
manufacturer/tradename
Excel Scientific TTS-100
manufacturer/tradename
Diversified Biotech T-TOPS-100
manufacturer/tradename
Diversified Biotech PSEAL-108C
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
packaging
pack of 50
packaging
pkg of 100 ea
packaging
pk of 100 ×
packaging
pack of 1 ea
feature
prevents well-to-well contamination, sample spillage and evaporation
feature
-
feature
prevents well-to-well contamination, sample spillage and evaporation, piercable
feature
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)