Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 180,00 zł |
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41122410
Manufacturer/tradename:
Diversified Biotech PSEAL-108C
Sterility:
non-sterile
Packaging:
pack of 1 ea
180,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócsterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech PSEAL-108C
L × W
12.5 in. × 32.9 in.
color
clear
General description
A sealing tape designed specifically for petri dishes and multiwell plates.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Conforms to the irregular perimeter to seal cover to plate, reduce evaporation, and prevent contamination. Waterproof, chemically resistant, and suitable for continuous use from −40 to +77°C. Heavy 5-mil (0.125 mm) thickness resists abrasion and puncture.
Legal Information
Petri-Seal is a trademark of C.B.S. Scientific Co.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description clear, L × W 12.5 in. × 32.9 in. | description blue, L × W 12.5 in. × 32.9 in. | description yellow, L × W 12.5 in. × 32.9 in. | description red, L × W 12.5 in. × 32.9 in. |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility - |
| manufacturer/tradename Diversified Biotech PSEAL-108C | manufacturer/tradename Diversified Biotech PSEAL-108B | manufacturer/tradename Diversified Biotech PSEAL-108Y | manufacturer/tradename Diversified Biotech PSEAL-108R |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| L × W 12.5 in. × 32.9 in. | L × W 12.5 in. × 32.9 in. | L × W 12.5 in. × 32.9 in. | L × W 12.5 in. × 32.9 in. |
| color clear | color blue | color yellow | color red |
Still not finding the right product?
Explore all of our products under Petri-Seal™ adhesive sealing film
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?