Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
Diversified Biotech CSEAL-75Y
W × l:
0.75 in. × 108 ft
Packaging:
pack of 1 ea
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócpackaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech CSEAL-75Y
W × L
0.75 in. × 108 ft
color
yellow
General description
ContainerSEAL is a sealing tape designed for the vast range of bottles and containers used in laboratory applications. Its unique elastomeric characteristics allow it to stretch and conform around the irregular perimeters of containers lids and covers to form a tight seal. Container Seal will block water vapor loss, reduce the rate of gas exchange and prevent the entry of contaminants.
Ideally sized (0.75" wide), the tape can be easily removed from most surfaces without leaving an adhesive residue. Available in a variety of colors.
Ideally sized (0.75" wide), the tape can be easily removed from most surfaces without leaving an adhesive residue. Available in a variety of colors.
Legal Information
ContainerSEAL is a registered trademark of Diversified Biotech
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description W × L 0.75 in. × 108 ft, yellow | description W × L 0.75 in. × 108 ft, red | description W × L 0.75 in. × 108 ft, white | description W × L 1 in. × 108 ft, white |
| manufacturer/tradename Diversified Biotech CSEAL-75Y | manufacturer/tradename Diversified Biotech CSEAL-75R | manufacturer/tradename Diversified Biotech CSEAL-75W | manufacturer/tradename Diversified Biotech CSEAL-XLW |
| W × L 0.75 in. × 108 ft | W × L 0.75 in. × 108 ft | W × L 0.75 in. × 108 ft | W × L 1 in. × 108 ft |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| color yellow | color red | color white | color white |
Still not finding the right product?
Explore all of our products under ContainerSEAL® sealing tape
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?