Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
blue , acrylonitrile-butadiene-styrene (ABS)
feature
Holds 50 x slides 76x26 mm
packaging
pack of 12 ea
manufacturer/tradename
Ted Pella 2304-4-CS
L × W × H
164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.)
General description
Features and Benefits
- Numbered index sheet in lid for quick identification
- Foam liner
- Stackable
- ABS-durable/impact resistant
- Hinged lid
- Unique sliding compression latch
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description blue, Holds 50 x slides 76x26 mm | description white, Holds 50 x slides 76x26 mm | description green, Holds 50 x slides 76x26 mm | description yellow, Holds 50 x slides 76x26 mm |
| feature Holds 50 x slides 76x26 mm | feature Holds 50 x slides 76x26 mm | feature Holds 50 x slides 76x26 mm | feature Holds 50 x slides 76x26 mm |
| manufacturer/tradename Ted Pella 2304-4-CS | manufacturer/tradename Ted Pella 2304-1-CS | manufacturer/tradename Ted Pella 2304-3-CS | manufacturer/tradename Ted Pella 2304-5-CS |
| material acrylonitrile-butadiene-styrene (ABS) , blue | material white , acrylonitrile-butadiene-styrene (ABS) | material acrylonitrile-butadiene-styrene (ABS) , green | material acrylonitrile-butadiene-styrene (ABS) , yellow |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea |
| L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) | L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) | L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) | L × W × H 164 mm (6.46 in.) × 110 mm (4.33 in.) × 32 mm (1.26 in.) |
Still not finding the right product?
Explore all of our products under PELCO® Sliding Latch Microslide Storage Boxes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?