Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
clear polycarbonate
Sterility:
non-sterile
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
clear polycarbonate
sterility
non-sterile
feature
lid
packaging
pack of 6 ea
manufacturer/tradename
Scaffdex C00007N
size
12 well
application(s)
3D cell culture
General description
CellCrown is a disposable insert, which immobilizes a sample in its place on a 6, 12, 24, 48, and 96-well plate. Fibers, foils, membranes, textiles, tissue and block like samples can be fixed with CellCrown in a firm, adjusted position in the well. The sample can be left in the well during the change of media by means of a pipette between the wall of the well and the CellCrown. The lid of the well plate can be closed while CellCrown is used. The insert can be removed from the well plate and unloaded after cell culturing without damaging the sample. CellCrown is made of medical grade polycarbonate and the product range includes sterile products ready to use as well as non-sterile products to be sterilized by the user. Sterilization can be by gamma or e-beam radiation, autoclaving, ethylene oxide gas and plasma sterilization.
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Sterilization Conditions:
Temperature.......121 °C / 250 °F
Time..................15 min.
Drying time.........15 min.
Legal Information
CellCrown is a trademark of Scaffdex Oy
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 12 well plate inserts, non-sterile | description 24 well plate inserts, sterile | description 6 well plate inserts, sterile | description 12 well plate inserts, sterile |
| manufacturer/tradename Scaffdex C00007N | manufacturer/tradename Scaffdex C00006S | manufacturer/tradename Scaffdex C00003S | manufacturer/tradename Scaffdex C00007S |
| sterility non-sterile | sterility sterile; γ-irradiated | sterility sterile | sterility sterile, sterile; γ-irradiated |
| material clear polycarbonate | material clear polycarbonate | material clear wells, clear polycarbonate | material polycarbonate , clear polycarbonate |
| size 12 well | size 24 well | size 6 wells | size 12 well |
| feature lid | feature lid | feature - | feature lid |
| packaging pack of 6 ea | packaging pack of 12 ea | packaging pack of 3 ea | packaging pack of 6 ea |
Still not finding the right product?
Explore all of our products under CellCrown™ inserts
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?