Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41100000
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócsterility
γ-irradiated
manufacturer/tradename
IBI ACCFL0001
General description
The IBI 3-channel Flow Cell (formerly Stovall 3-channel Flow Cell) is designed as a one-time use, affordable, sterile 3-channel flow cell apparatus for the creation of biofilms. This device allows for real-time, direct, non-destructive, on-line examination of biofilms with either a traditional transmission light microscope or a scanning confocal laser microscope which will produce a 3-D image of the biofilm.
The 3-channel flow cell is gamma irradiated and comes complete with bubble trap (ACCFL0002) and peristaltic pump tubing and connective tubing. The 3-channel flow cell contains a #1 Glass cover slip. The underside of this cover slip is where the biofilm will attach and grow. When the experiment is complete, the cover slip is cut using a hand-held glass cutter (ACCFL0014) and removed along with the biofilm.
The 3-channel flow cell is gamma irradiated and comes complete with bubble trap (ACCFL0002) and peristaltic pump tubing and connective tubing. The 3-channel flow cell contains a #1 Glass cover slip. The underside of this cover slip is where the biofilm will attach and grow. When the experiment is complete, the cover slip is cut using a hand-held glass cutter (ACCFL0014) and removed along with the biofilm.
Features and Benefits
- The 3 separate channels allow for the simultaneous growth of 3 individual biofilms in the same cell.
- 3-channel cell only – No tubing included.
- Cell comes sterilized (gamma irradiated) and ready for use.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Cell Only, γ-irradiated, Sealed | description Complete Assembly, γ-irradiated | description Cell Only, Glass Cover Slip, sterile; γ-irradiated | description Complete Assembly, Glass Cover Slip, sterile; γ-irradiated |
| sterility γ-irradiated | sterility γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| manufacturer/tradename IBI ACCFL0001 | manufacturer/tradename IBI FLCAS0001 | manufacturer/tradename IBI CFCAS0003 | manufacturer/tradename IBI CFCAS0001 |
Still not finding the right product?
Explore all of our products under 3-channel flow cell
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?