Przejdź do
Z741749
3-channel flow cell
Complete Assembly, γ-irradiated
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1120,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41100000
1120,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
sterility
γ-irradiated
manufacturer/tradename
IBI FLCAS0001
General description
The IBI 3-channel Flow Cell (formerly Stovall 3-channel Flow Cell) is designed as a one-time use, affordable, sterile 3-channel flow cell apparatus for the creation of biofilms. This device allows for real-time, direct, non-destructive, on-line examination of biofilms with either a traditional transmission light microscope or a scanning confocal laser microscope which will produce a 3-D image of the biofilm.
The 3-channel flow cell is gamma irradiated and comes complete with bubble trap (ACCFL0002) and peristaltic pump tubing and connective tubing. The 3-channel flow cell contains a #1 Glass cover slip. The underside of this cover slip is where the biofilm will attach and grow. When the experiment is complete, the cover slip is cut using a hand-held glass cutter (ACCFL0014) and removed along with the biofilm.
Features and Benefits
  • The 3 separate channels allow for the simultaneous growth of 3 individual biofilms in the same cell.
  • Luer connectors in the effluent tubing (downstream from the cell) allow for collection of effluent for further analysis.
  • 3-channel full assembly measures 91 inches long including cell and tubing.
  • Included bubble trap captures air bubbles before they reach the cell and also smooth peristaltic pump pulsations.
  • System comes sterilized (gamma irradiated) and ready for use.
  • Culture media bottles and peristaltic pump purchased separately.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z741750Z741751Z741764
description
Complete Assembly, γ-irradiated
description
Cell Only, γ-irradiated, Sealed
description
-
description
Complete Assembly, Plastic Cover Slip, sterile; γ-irradiated
sterility
γ-irradiated
sterility
γ-irradiated
sterility
sterile; irradiated
sterility
sterile; γ-irradiated
manufacturer/tradename
IBI FLCAS0001
manufacturer/tradename
IBI ACCFL0001
manufacturer/tradename
IBI ACCFL0002
manufacturer/tradename
IBI CFCAS0002
Still not finding the right product?
Explore all of our products under 3-channel flow cell
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)