Przejdź do
Z741649
Sartorius pipette tips
volume range 100-5000 μL, Standard, rack, sterile
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121600
eCl@ss:
32079201
Volume:
100—5000 μL
Sterility:
sterile
Feature:
DNase free, RNase free, barrier: no, endotoxin free
Packaging:
pack of 50 ea
Manufacturer/tradename:
Sartorius 780305
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
colorless
sterility
sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
packaging
pack of 50 ea
manufacturer/tradename
Sartorius 780305
maximum volume
5 mL
volume range
100-5000 μL
volume
100—5000 μL
General description
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
Browse all pipette tips
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z741648Z741651Z757748
description
volume range 100-5000 μL, Standard, rack, sterile
description
volume range 100-5000 μL, Standard, rack, non-sterile
description
volume range 500-10000 μL, Standard, bulk, non-sterile
description
volume range 10-1000 μL, standard, refill, sterile
feature
endotoxin free, RNase free, barrier: no, DNase free
feature
DNase free, RNase free, endotoxin free, barrier: no
feature
RNase free, barrier: no, endotoxin free, DNase free
feature
endotoxin free, DNase free, barrier: no, RNase free
volume
100—5000 μL
volume
100—5000 μL
volume
500—10000 μL
volume
10—1000 μL
sterility
sterile
sterility
non-sterile
sterility
non-sterile
sterility
sterile
manufacturer/tradename
Sartorius 780305
manufacturer/tradename
Sartorius 780304
manufacturer/tradename
Sartorius 780310
manufacturer/tradename
Sartorius 791003
material
colorless
material
colorless
material
colorless , colorless tip
material
colorless
packaging
pack of 50 ea
packaging
pack of 50 ea
packaging
pack of 100 ea
packaging
pack of 96 ea (Racked)
Still not finding the right product?
Explore all of our products under Sartorius pipette tips
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)