Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
50 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 78,40 zł |
Informacje o tej pozycji
78,40 zł
material
colorless
sterility
non-sterile
feature
DNase free, RNase free, barrier: no, endotoxin free
packaging
pack of 50 ea
manufacturer/tradename
Sartorius 780304
maximum volume
5 mL
volume range
100-5000 μL
volume
100—5000 μL
General description
Optifit Tips are available in various packaging types for all your needs:
- Single Tray racks – purity certified tip racks
- Refill Packs - purity certified and ecological refill packages
- Refill Towers - space-saving and ecological refill packages
- Flexibulk boxes - space-saving, ecological and economical bulk boxes
- Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
Other Notes
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume range 100-5000 μL, Standard, rack, non-sterile | description volume range 500-10000 μL, Standard, bulk, non-sterile | description volume range 100-5000 μL, standard, bulk packed, non-sterile | description volume range 100-5000 μL, Standard, bulk, non-sterile |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| volume 100—5000 μL | volume 500—10000 μL | volume 100—5000 μL | volume 100—5000 μL |
| manufacturer/tradename Sartorius 780304 | manufacturer/tradename Sartorius 780310 | manufacturer/tradename Sartorius 780300 | manufacturer/tradename Sartorius 780308 |
| feature barrier: no, RNase free, DNase free, endotoxin free | feature barrier: no, endotoxin free, RNase free, DNase free | feature DNase free, RNase free, endotoxin free, barrier: no | feature endotoxin free, DNase free, barrier: no, RNase free |
| material colorless | material colorless tip | material colorless | material colorless |
| packaging pack of 50 ea | packaging pack of 100 ea | packaging pack of 100 ea (Color coded tip trays) | packaging pack of 1000 ea |
Still not finding the right product?
Explore all of our products under Sartorius pipette tips
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?