Zmień widok
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
50 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 221,00 zł |
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Material:
polyethylene
221,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
polyethylene
packaging
package of 50 (individually wrapped)
manufacturer/tradename
Sartorius 721016
diam.
5.33 mm
General description
The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINEmLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description diam. 5.33 mm, polyethylene | description diam. 6.73 mm, polyethylene | description diam. 3.15 mm, polyethylene | description diam. 6.73 mm, polyethylene |
| material polyethylene | material polyethylene | material polyethylene | material polyethylene |
| diam. 5.33 mm | diam. 6.73 mm | diam. 3.15 mm | diam. 6.73 mm |
| manufacturer/tradename Sartorius 721016 | manufacturer/tradename Sartorius 721015 | manufacturer/tradename Sartorius 721017 | manufacturer/tradename Sartorius 721005 |
| packaging package of 50 (individually wrapped) | packaging package of 50 (individually wrapped) | packaging package of 50 (individually wrapped) | packaging package of 50 (individually wrapped) |
Still not finding the right product?
Explore all of our products under Plus Safe-Cone Filter
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?