Przejdź do
Z741602
Standard Safe-Cone Filter
diam. 6.73 mm, polyethylene
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
50 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
180,00 zł
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Material:
polyethylene
180,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
polyethylene
packaging
package of 50 (individually wrapped)
manufacturer/tradename
Sartorius 721005
diam.
6.73 mm
General description
The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette. Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Plus Filter is recommended for, but need to be changed more frequently. Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology. The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration.To ensure safety of the user, forceps should be used to change the filter. The mLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter. The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette. Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Plus Filter is recommended for, but need to be changed more frequently. Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology. The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration.To ensure safety of the user, forceps should be used to change the filter. The mLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.
Legal Information
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z741603Z741605Z741606
description
diam. 6.73 mm, polyethylene
description
diam. 5.33 mm, polyethylene
description
diam. 3.15 mm, polyethylene
description
diam. 2.51 mm, polyethylene
material
polyethylene
material
polyethylene
material
polyethylene
material
polyethylene
packaging
package of 50 (individually wrapped)
packaging
package of 50 (individually wrapped)
packaging
package of 50 (individually wrapped)
packaging
package of 50 (individually wrapped)
manufacturer/tradename
Sartorius 721005
manufacturer/tradename
Sartorius 721006
manufacturer/tradename
Sartorius 721007
manufacturer/tradename
Sartorius 721008
diam.
6.73 mm
diam.
5.33 mm
diam.
3.15 mm
diam.
2.51 mm
Still not finding the right product?
Explore all of our products under Standard Safe-Cone Filter
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)