Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
500 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1250,00 zł |
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
polyethylene cap, polypropylene container
1250,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomócmaterial
polyethylene cap, polypropylene container
packaging
pack of 500 ea
manufacturer/tradename
Simport C577-20W
size
20 mL
General description
Chemically resistant and shatterproof, The SecurTainer III Specimen Containers are supplied with a leakproof screw cap, particularly important when transporting hazardous material. Ideal for transport and storage of urine, sputum and most liquids or particulate samples.
Containers are uniquely stackable, shatter resistant and are manufactured from virgin, high-clarity polypropylene. Available in many sizes from 20 to 120 ml. Containers and caps packaged separately in bags of 100. Non sterile. Functional temperature range: -90 °C to +100 °C.
Containers are uniquely stackable, shatter resistant and are manufactured from virgin, high-clarity polypropylene. Available in many sizes from 20 to 120 ml. Containers and caps packaged separately in bags of 100. Non sterile. Functional temperature range: -90 °C to +100 °C.
Legal Information
SecurTainer is a trademark of Simport
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size 20 mL | description size 40 mL | description size 90 mL | description size 120 mL |
| material polyethylene cap, polypropylene container | material polyethylene cap, polypropylene container | material polypropylene container, polyethylene cap | material polypropylene container, polyethylene cap |
| size 20 mL | size 40 mL | size 90 mL | size 120 mL |
| manufacturer/tradename Simport C577-20W | manufacturer/tradename Simport C577-40W | manufacturer/tradename Simport C577-90W | manufacturer/tradename Simport C577-120W |
| packaging pack of 500 ea | packaging pack of 500 ea | packaging pack of 400 ea | packaging pack of 300 ea |
Still not finding the right product?
Explore all of our products under SecurTainer™ III specimen containers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?