Przejdź do
Z740375
Eco-Friendly SpecTainer™, tamper evident, biodegradable
size 60 mL, sterile
Synonim(y):
Simport Plastics C566-60AQSECO, bidegradable sample storage, sample receptacle, sample storage, secure sample container
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
500 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
3380,00 zł
2873,00 zł
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
2873,00 zł
Cena katalogowa3380,00 złZaoszczędź 15%
Promocja dostępna wyłącznie online
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
polyethylene cap, polypropylene container
sterility
sterile
packaging
pack of 500 ea
manufacturer/tradename
Simport C566-60AQSECO
size
60 mL
General description
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.
The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.
Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
Legal Information
SpecTainer is a trademark of Simport
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z740377Z740376Z740379
description
size 60 mL, sterile
description
size 90 mL, sterile
description
size 60 mL, non-sterile
description
size 120 mL, sterile
manufacturer/tradename
Simport C566-60AQSECO
manufacturer/tradename
Simport C566-90AQSECO
manufacturer/tradename
Simport C566-60DOECO
manufacturer/tradename
Simport C566-120AQSECO
material
polypropylene container, polyethylene cap
material
polypropylene container, polyethylene cap
material
polyethylene cap, polypropylene container
material
polypropylene container, polyethylene cap
packaging
pack of 500 ea
packaging
pack of 400 ea
packaging
pack of 500 ea
packaging
pack of 300 ea
size
60 mL
size
90 mL
size
60 mL
size
120 mL
sterility
sterile
sterility
sterile
sterility
non-sterile
sterility
sterile
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)