Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 23,00 zł |
Informacje o tej pozycji
23,00 zł
material
colorless polypropylene box, lid, and holder (includes a colored tip-tray)
Quality Segment
sterility
non-sterile
feature
autoclavable (121°C, 2 bar), Holds <50 μl tips, stackable
packaging
box of 1
manufacturer/tradename
BRAND 732990
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description with gray tip-tray for pipette and filter tips up to 50 μL | description with green tip-tray for 300 μL tips | description with yellow tip-tray for 200 μL tips | description with blue tip-tray for 1000 μL tips |
| feature autoclavable (121°C, 2 bar), stackable, Holds <50 μl tips | feature Holds <300 μl tips, stackable, autoclavable (121°C, 2 bar) | feature autoclavable (121°C, 2 bar), Holds <200 μl tips, stackable | feature Holds <1000 μl tips, autoclavable (121°C, 2 bar), stackable |
| manufacturer/tradename BRAND 732990 | manufacturer/tradename BRAND 732994 | manufacturer/tradename BRAND 732992 | manufacturer/tradename BRAND 732996 |
| material colorless polypropylene box, lid, and holder (includes a colored tip-tray) | material colorless polypropylene box, lid, and holder (includes a colored tip-tray) | material colorless polypropylene box, lid, and holder (includes a colored tip-tray) | material colorless polypropylene box, lid, and holder (includes a colored tip-tray) |
| packaging box of 1 | packaging box of 1 | packaging box of 1 | packaging box of 1 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| Quality Level 200 | Quality Level - | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under BRAND® TipBox, empty
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?