Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
960 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 434,00 zł |
Informacje o tej pozycji
434,00 zł
material
colorless , colorless polypropylene tip and filter element (polyethylene)
sterility
sterile; autoclaved
feature
RNase and DNase free, endotoxin free, filter, BIO-CERT®
packaging
pack of 960 ea (10 racks of 96)
manufacturer/tradename
BRAND 732630
maximum volume
100 μL
volume
5-100 μL
General description
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. These filters function with consistent reliability. On the other hand, liquids can pass very slowly should they accidentally contact the filter.
Since the filter does not swell as in the case of self-sealing filter tips, the sample can be recovered from the filter by simply actuating the pipette′s blow-out function, or by centrifugation if necessary. This is clearly an important advantage of non-self-sealing filters, especially when working with valuable samples. As an added advantage, the absence of filter additives protects samples from this source of contamination.
NEW! TipRack, non-sterile and sterile, BIO-CERT®
Space-saving refill unit for TipBox. Protected in an environmentally compatible packaging of recyclable PET. Compared to the filled TipBoxes, the amount of waste with the new refill units is reduced by 20%. All pipette tips and filter tips up to 1000 μL are free of DNA, RNases, endotoxins and ATP. TipRacks in BIO-CERT® quality are sterile according to ISO 11 137 and the AAMI guidelines, a SAL of 10-6 is obtained.
These racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilized TipBox. All tip-trays are printed on one side with information on the contents.
Features and Benefits
- Cleanroom quality for contamination-free analyses.
- Perfect fit for the Transferpette® micropipette and many other pipettes.
- The tip system for all requirements.
Other Notes
Legal Information
1 of 1
Ta pozycja | ||
|---|---|---|
| description volume 5-100 μL, BIO-CERT® | description volume 50-1000 μL, BIO-CERT® | description volume 2-20 μL, BIO-CERT® |
| volume 5-100 μL | volume 50-1000 μL | volume 2-20 μL |
| manufacturer/tradename BRAND 732630 | manufacturer/tradename BRAND 732634 | manufacturer/tradename BRAND 732628 |
| sterility sterile; autoclaved | sterility sterile; autoclaved | sterility sterile; autoclaved |
| feature RNase and DNase free, filter, endotoxin free, BIO-CERT® | feature RNase and DNase free, endotoxin free, BIO-CERT® | feature RNase and DNase free, filter, endotoxin free, BIO-CERT® |
| material colorless , colorless polypropylene tip and filter element (polyethylene) | material colorless polypropylene tip and filter element (polyethylene) | material colorless polypropylene tip and filter element (polyethylene) |
| packaging pack of 960 ea (10 racks of 96) | packaging pack of 960 ea (10 racks of 96) | packaging pack of 960 ea (10 racks of 96) |
Still not finding the right product?
Explore all of our products under BRAND® filter tips, racked, TipRack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?