Przejdź do
Z740019
BRAND® filter tips, racked, TipRack
volume 2-20 μL, BIO-CERT®
Synonim(y):
filter pipette tips, pipette filter tips, pipette tips with filters, racked filter tips
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
960 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
423,00 zł
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121601
eCl@ss:
32079202
Volume:
2-20 μL
Sterility:
sterile; autoclaved
Feature:
RNase and DNase free, endotoxin free, filter, BIO-CERT®
Packaging:
pack of 960 ea (10 racks of 96)
Manufacturer/tradename:
BRAND 732628
423,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
material
colorless , colorless polypropylene tip and filter element (polyethylene)
sterility
sterile; autoclaved
feature
RNase and DNase free, endotoxin free, filter, BIO-CERT®
packaging
pack of 960 ea (10 racks of 96)
manufacturer/tradename
BRAND 732628
maximum volume
20 μL
volume
2-20 μL
General description
Pipette tips and filter tips are manufactured by BRAND in a cleanroom under the most modern production conditions, and are automatically rack packed and packaged to ensure that the tips are of consistently high quality.
Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipette shaft. These filters function with consistent reliability. On the other hand, liquids can pass very slowly should they accidentally contact the filter.
Since the filter does not swell as in the case of self-sealing filter tips, the sample can be recovered from the filter by simply actuating the pipette′s blow-out function, or by centrifugation if necessary. This is clearly an important advantage of non-self-sealing filters, especially when working with valuable samples. As an added advantage, the absence of filter additives protects samples from this source of contamination.
NEW! TipRack, non-sterile and sterile, BIO-CERT®
Space-saving refill unit for TipBox. Protected in an environmentally compatible packaging of recyclable PET. Compared to the filled TipBoxes, the amount of waste with the new refill units is reduced by 20%. All pipette tips and filter tips up to 1000 μL are free of DNA, RNases, endotoxins and ATP. TipRacks in BIO-CERT® quality are sterile according to ISO 11 137 and the AAMI guidelines, a SAL of 10-6 is obtained.
These racks are supplied with a transfer aid that enables simple, contamination-free transfer into a previously sterilized TipBox. All tip-trays are printed on one side with information on the contents.
Features and Benefits
  • Cleanroom quality for contamination-free analyses.
  • Perfect fit for the Transferpette® micropipette and many other pipettes.
  • The tip system for all requirements.
Other Notes
BR732628 (India)
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z740065Z740017Z740015
description
volume 2-20 μL, BIO-CERT®
description
volume 50-1000 μL, BIO-CERT®
description
volume 1-20 μL, BIO-CERT®
description
volume 0.5-10 μL, BIO-CERT®
volume
2-20 μL
volume
50-1000 μL
volume
1-20 μL
volume
0.5-10 μL
manufacturer/tradename
BRAND 732628
manufacturer/tradename
BRAND 732634
manufacturer/tradename
BRAND 732626
manufacturer/tradename
BRAND 732624
sterility
sterile; autoclaved
sterility
sterile; autoclaved
sterility
sterile; autoclaved
sterility
sterile; autoclaved
feature
RNase and DNase free, filter, BIO-CERT®, endotoxin free
feature
RNase and DNase free, endotoxin free, BIO-CERT®
feature
endotoxin free, filter, BIO-CERT®, RNase and DNase free
feature
filter, RNase and DNase free, endotoxin free, BIO-CERT®
material
colorless polypropylene tip and filter element (polyethylene)
material
colorless polypropylene tip and filter element (polyethylene)
material
colorless polypropylene tip and filter element (polyethylene)
material
colorless polypropylene tip and filter element (polyethylene)
packaging
pack of 960 ea (10 racks of 96)
packaging
pack of 960 ea (10 racks of 96)
packaging
pack of 960 ea (10 racks of 96)
packaging
pack of 960 ea (10 racks of 96)
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)