Przejdź do
Z720380
Nalgene® centrifuge tubes, Oak Ridge style 3115
nominal capacity 30 mL
Synonim(y):
Nalgene centrifuge tube, plastic centrifuge tube, Nalgene 3115-0030, sample collection tube, sample receptacle, sample tube
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121703
eCl@ss:
32040302
Material:
polysulfone , round bottom tube, screw top cap (Linerless), transparent tube
Packaging:
pack of 10, case of 100
Feature:
RCF 50,000 × g
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomóc
material
polysulfone , round bottom tube, screw top cap (Linerless), transparent tube
feature
RCF 50,000 × g
packaging
pack of 10, case of 100
manufacturer/tradename
Nalgene 3115-0030
nominal capacity
30 mL
General description
Easily view contents when using these strong, transparent Nalgene®polysulfone tubes featuring enhanced resistance to acids and bases. Can be used up to 50,000 x g in refrigerated or non-refrigerated centrifuges.
Features and Benefits
  • Round bottoms to fit high speed rotors.
  • Transparent with straw-colored cast.
  • Comes with polypropylene linerless closures—easy to clean and provides leakproof† protection under normal conditions up to 10,000 x g.
  • When equipped with a Nalgene sealing cap (sold separately), 30mL and 50mL tubes can be spun up to 50,000 x g.
  • Can be used in refrigerated or non-refrigerated centrifuges.
  • Tubes must be filled to at least 80% capacity for proper performance.
  • Autoclavable/Leakproof† when used with supplied Nalgene closure.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z720399T1293T1793
description
nominal capacity 30 mL
description
nominal capacity 50 mL
description
nominal capacity 30 mL
description
nominal capacity 30 mL
manufacturer/tradename
Nalgene 3115-0030
manufacturer/tradename
Nalgene 3115-0050
manufacturer/tradename
Nalgene® 3119-0030
manufacturer/tradename
Nalgene® 3118-0030
material
transparent tube, screw top cap (Linerless), round bottom tube, polysulfone
material
screw top cap (Linerless), polysulfone , round bottom tube, transparent tube
material
round bottom tube, screw top cap (Linerless), polypropylene screw closure, polypropylene copolymer
material
polypropylene screw closure, transparent polycarbonate , round bottom tube, screw top cap
feature
RCF 50,000 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
packaging
case of 100, pack of 10
packaging
pack of 10, case of 100
packaging
pack of 10 ea
packaging
pack of 10 ea
nominal capacity
30 mL
nominal capacity
50 mL
nominal capacity
30 mL
nominal capacity
30 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)