Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2650,00 zł |
Informacje o tej pozycji
2650,00 zł
feature
electronic: no, single-channel
packaging
pack of 1 ea, pkg of 1 ea
manufacturer/tradename
Nichiryo 00-NPL02-1000
availability
not available in (Not available in France)
volume
100-1000 μL
General description
Time and money spent on the calibration of pipettes is now greatly reduced!
- UV resistant
- Solvent resistant
- Fully autoclavable (121°C for 20 minutes)
- One-touch calibration
- Ceramic pistons
- Filtered nozzles
- Easy, convenient locking lever
- Ergonomic design
- Single channel, manual, continuously adjustable volume
- Advanced durability against organic solvent dispensing
- PTFE (Fluorine resin) in the air tight chamber of the instrument
- A Perfluoro rubber O-ring with low cubical expansion in the airtight parts
- Plunger made from a material resistant to organic solvents
- An alloy spring to avoid metal corrosion by organic solvents
- Quick and easy to use
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 100-1000 μL | description volume 10-100 μL | description volume 1000-5000 μL | description volume 0.1-2.0 μL |
| feature single-channel, electronic: no | feature electronic: no, single-channel | feature single-channel, electronic: no | feature single-channel, electronic: no |
| volume 100-1000 μL | volume 10-100 μL | volume 1000-5000 μL | volume 0.1-2.0 μL |
| manufacturer/tradename Nichiryo 00-NPL02-1000 | manufacturer/tradename Nichiryo 00-NPL02-100 | manufacturer/tradename Nichiryo 00-NPL02-5000 | manufacturer/tradename Nichiryo 00-NPL02-2 |
| packaging pkg of 1 ea, pack of 1 ea | packaging pkg of 1 ea, pack of 1 ea | packaging pkg of 1 ea, pack of 1 ea | packaging pack of 1 ea, pkg of 1 ea |
| availability not available in (Not available in France) | availability not available in (Not available in France) | availability not available in (Not available in France) | availability not available in (Not available in France) |
Still not finding the right product?
Explore all of our products under Nichipet EX-Plus II pipette
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?