Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
6 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 498,00 zł 423,30 zł |
Informacje o tej pozycji
423,30 zł
Cena katalogowa498,00 złZaoszczędź 15%material
transparent polycaprolactone (PCL) , transparent wells
sterility
sterile; electron beam irradiated, sterile
packaging
pack of 6 ea
manufacturer/tradename
Nanofiber Solutions 60602
size
6 wells
pore size
700 nm (Fiber spacing)
suitability
suitable for (Tissue Engineering, Cancer Research)
application(s)
3D cell culture
binding type
(plasma surface treated)
General description
The nanofibers mimic the 3D topography in vivo, and can be easily coated with ECM proteins for cancer research. The cells can be simply removed for protein or gene analysis.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining.
Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for 6 well plates | description for 6 well plates | description size 6 wells | description size 6 wells |
| binding type (plasma surface treated) | binding type (plasma surface treated) | binding type non-treated surface | binding type non-treated surface |
| manufacturer/tradename Nanofiber Solutions 60602 | manufacturer/tradename Nanofiber Solutions 60601 | manufacturer/tradename Nanofiber Solutions 601 | manufacturer/tradename Nanofiber Solutions 602 |
| pore size 700 nm (Fiber spacing) | pore size 700 nm (Fiber spacing) | pore size - | pore size - |
| sterility sterile, sterile; electron beam irradiated | sterility sterile; electron beam irradiated | sterility sterile | sterility sterile |
| material transparent polycaprolactone (PCL) , transparent wells | material flat clear wells, (polycaprolactone (PCL)), transparent polycaprolactone (PCL) | material (polycaprolactone (PCL)), flat bottom clear wells, transparent polycaprolactone (PCL) | material flat bottom clear wells, (polycaprolactone (PCL)) |
| suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue Engineering, Cancer Research) | suitability suitable for (Tissue engineering, cancer research) |
Still not finding the right product?
Explore all of our products under Nanofiber multiwell plate inserts, with aligned nanofibers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?