Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 451,00 zł |
Informacje o tej pozycji
451,00 zł
material
(polycaprolactone (PCL)), flat bottom clear wells
sterility
sterile
feature
lid
manufacturer/tradename
Nanofiber Solutions 602
technique(s)
cell culture: suitable
size
6 wells
suitability
suitable for (Tissue engineering, cancer research)
application(s)
3D cell culture
binding type
non-treated surface
General description
The cell culture dishes, plates and inserts are available with either randomly oriented fibers or aligned fibers.
The nanofibers have higher expansion rates, better differentiation control, and has a synthetic surface with no animal contamination, ideal for stem cells research and immunohistochemistry staining. Nanofiber scaffolds can be easily shaped or sized, made from nearly any synthetic or natural polymer for customized tissue engineering. They can then be implanted in vivo or used in vitro. The standard plate dimensions available will work with current automated equipment for high throughput drug discovery needs.
For fluorescent imaging applications, we recommend using the Nanofiber Solutions chamber slides.
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size 6 wells | description size 384 wells | description size 96 wells | description size 24 wells |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| feature lid | feature lid | feature lid | feature lid |
| manufacturer/tradename Nanofiber Solutions 602 | manufacturer/tradename Nanofiber Solutions 38402 | manufacturer/tradename Nanofiber Solutions 9602 | manufacturer/tradename Nanofiber Solutions 2402 |
| binding type non-treated surface | binding type non-treated surface | binding type non-treated surface | binding type non-treated surface |
| material (polycaprolactone (PCL)), flat bottom clear wells | material flat bottom clear wells, (polycaprolactone (PCL)), transparent polycaprolactone (PCL) | material flat bottom clear wells, transparent polycaprolactone (PCL) , (polycaprolactone (PCL)) | material transparent polycaprolactone (PCL) , flat clear wells, (polycaprolactone (PCL)) |
| size 6 wells | size 384 wells | size 96 wells | size 24 wells |
Still not finding the right product?
Explore all of our products under Nanofiber multiwell plate, with aligned nanofibers
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?