Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
10 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3550,00 zł |
Informacje o tej pozycji
3550,00 zł
material
borosilicate glass , flat bottom flask
joint
joint ST/NS neck45 mm I.D.
sterility
non-sterile
packaging
pkg of 1 ea, pkg of 10 ea
manufacturer/tradename
DWK Life Sciences DURAN® 212045458
availability
not available in USA and Canada
capacity
1000 mL
General description
Manufactured from Duran borosilicate glass type 3.3, the flasks have maximum chemical and thermal resistance and meet ISO 6556 standard.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 1000 mL | description capacity 100 mL | description capacity 500 mL | description capacity 250 mL |
| capacity 1000 mL | capacity 100 mL | capacity 500 mL | capacity 250 mL |
| manufacturer/tradename DWK Life Sciences DURAN® 212045458 | manufacturer/tradename DWK Life Sciences DURAN® 212042452 | manufacturer/tradename DWK Life Sciences DURAN® 212044453 | manufacturer/tradename DWK Life Sciences DURAN® 212043654 |
| material borosilicate glass , flat bottom flask | material borosilicate glass , flat bottom flask | material flat bottom flask, borosilicate glass | material borosilicate glass , flat bottom flask |
| joint joint ST/NS neck45 mm I.D. | joint joint ST/NS neck24 mm | joint joint ST/NS neck34 mm I.D. | joint joint ST/NS neck34 mm I.D. |
| packaging pkg of 10 ea, pkg of 1 ea | packaging pkg of 1 ea, pkg of 10 ea | packaging pkg of 1 ea, pkg of 10 ea | packaging pkg of 10 ea, pkg of 1 ea |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
Still not finding the right product?
Explore all of our products under Duran® filtering flask, with Keck® assembly set
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?