Przejdź do
Z688533
Microscope slide-grids, disposable
use with dry or suspended specimens
Synonim(y):
Diversified Biotech GRID-1000, adhesive well sticker, gridded slide well stickers, sample isolation sticker, well stickers
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 pkg
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
180,00 zł
Informacje o tej pozycji
NACRES:
MA.01
UNSPSC Code:
41122600
Material:
polyester
W × l:
20 mm × 20 mm
Packaging:
pack of 40 ea
Manufacturer/tradename:
Diversified Biotech GRID-1000
180,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
polyester
packaging
pack of 40 ea
manufacturer/tradename
Diversified Biotech GRID-1000
technique(s)
cell culture: suitable
W × L
20 mm × 20 mm
General description
Convenient- Easy-to adhere stickers having side-by-side twin grids (20 mm x 20 mm each with 1 mm line spacing) for microscope slides. Count or measure cells and estimate the size or length of microscopic or semi-micro objects
Versatile- Compatible with different styles of microscopes (conventional, inverted or dissecting). Virtually all cells and specimens placed on top of Slide-Grids can be counted and/or sized.
Dry or Wet- Use with dry specimens or suspended specimens such as cells held in buffer under cover slips. Separate cover slips can be placed over each Slide-Grid.
Chemically Inert- Made of polyester with an acrylic adhesive. Slide-Grids are inert and stable under most laboratory conditions.
Cover slips not included
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z743543Z743544XX1007615
description
use with dry or suspended specimens
description
well diam. × depth 1 cm × 0.25 mm, pack of 25
description
well diam. × depth 1 cm × 0.50 mm, pack of 25
description
Accessories for filter holders for sample preparation.
W × L
20 mm × 20 mm
W × L
-
W × L
-
W × L
-
manufacturer/tradename
Diversified Biotech GRID-1000
manufacturer/tradename
Diversified Biotech MIWE-1000
manufacturer/tradename
Diversified Biotech MIWE-2000
manufacturer/tradename
-
material
polyester
material
-
material
-
material
glass fiber slide
packaging
pack of 40 ea
packaging
pack of 25
packaging
pack of 25
packaging
-
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)