Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
material
titanium (Ti-6AI-4V alloy)
packaging
pack of 1 ea
manufacturer/tradename
Bandelin 494
availability
available only in EU
diam. × L
13 mm × 126 mm
diam.
≥17 mm
volume
10-200 mL , HD 2070, 20-900 mL , HD 2200
General description
Using the product image (from left to right):
MS 72 micro tip
MS 73 micro tip
KE 76 tapered tip
VS 70 T extended probe
VS 200 T extended probe
TT 13 flat tip
TT 19 flat tip
TT 25 flat tip
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description VS 70 T extended probe for use with HD 2070 homogenizer and SG70G horn or HD 2200 homogenizer and SG 213G horn | description VS 200 T extended probe for use with HD 2070 homogenizer and SG70G horn or HD 2200 homogenizer and SG 213G horn | description TT 13 flat tip | description KE 76 tapered tip for use with HD 2070 homogenizer and SG70G horn or HD 2200 homogenizer and SG 213G horn |
| packaging pack of 1 ea | packaging - | packaging - | packaging pack of 1 ea |
| material titanium (Ti-6AI-4V alloy) | material titanium (Ti-6AI-4V alloy) | material titanium (Ti-6AI-4V alloy) | material titanium (Ti-6AI-4V alloy) |
| diam. ≥17 mm | diam. - | diam. 13 mm | diam. ≥8 mm |
| availability available only in EU | availability available only in EU | availability available only in EU | availability available only in EU |
| manufacturer/tradename Bandelin 494 | manufacturer/tradename Bandelin 500 | manufacturer/tradename Bandelin 497 | manufacturer/tradename Bandelin 530 |
| volume 10-200 mL , HD 2070, 20-900 mL , HD 2200 | volume - | volume 10-200 mL , HD 2070, 20-900 mL , HD 2200 | volume 5-100 mL , HD 2070, 10-350 mL , HD 2200 |
Still not finding the right product?
Explore all of our products under Bandelin SONOPULS ultrasonic homogenizer probes
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?