Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1000 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 132,00 zł |
Informacje o tej pozycji
132,00 zł
material
colorless
sterility
non-sterile
feature
barrier: no, low retention
packaging
pack of 1000 ea (bag)
manufacturer/tradename
Nichiryo BMTL-SS
availability
not available in (France)
maximum volume
10 μL
volume range
0.1-10 μL
volume
0.1—10 μL
Features and Benefits
- RNase and DNase free
- Fits most pipettes
- Premium packaging featuring stand-up and puncture-resistant bags
- Racked tips are offered in both hinged and lift-off tops
Other Notes
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume range 0.1-10 μL, elongated, bulk pack | description maximum volume 200 μL, universal, pack of 960 (10 racks of 96 tips), sterile | description volume range 1-10 mL, universal, bulk pack | description maximum volume 5 mL, universal, bulk pack |
| volume 0.1—10 μL | volume - | volume 1—10 mL | volume 5 mL |
| feature barrier: no, low retention | feature barrier: no | feature barrier: no, RNase free, DNase free | feature RNase free, DNase free, barrier: no |
| sterility non-sterile | sterility sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Nichiryo BMTL-SS | manufacturer/tradename Nichiryo BMT-SGRS | manufacturer/tradename Nichiryo BMT-Z | manufacturer/tradename Nichiryo BMT-XR |
| material colorless | material colorless | material colorless , clear tip | material colorless |
| packaging pack of 1000 ea (bag) | packaging pack of 960 (10 racks of 96 tips) | packaging pack of 200 ea (Bag) | packaging pack of 100 ea (Bag) |
Still not finding the right product?
Explore all of our products under Nichiryo pipette tips
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?