Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2380,00 zł |
Informacje o tej pozycji
2380,00 zł
material
polycarbonate housing, polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 100 ea
manufacturer/tradename
Sartorius VS0132
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
~2.2 mL
working volume
500 μL
pore size
50 kDa MWCO
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
1 of 1
Ta pozycja | ||
|---|---|---|
| description MWCO 50 kDa | description MWCO 10 kDa | description MWCO 50 kDa |
| manufacturer/tradename Sartorius VS0132 | manufacturer/tradename Sartorius VS0102 | manufacturer/tradename Sartorius VS0131 |
| parameter 15000 × g max. centrifugal force | parameter 15000 × g max. centrifugal force | parameter 15000 × g max. centrifugal force |
| pore size 50 kDa MWCO | pore size 10 kDa MWCO | pore size 50 kDa MWCO |
| sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material polyethersulfone membrane, polycarbonate housing | material polyethersulfone membrane, polycarbonate housing | material polycarbonate housing, polyethersulfone membrane |
| working volume 500 μL | working volume 500 μL | working volume 500 μL |
Still not finding the right product?
Explore all of our products under Vivaspin 500 centrifugal concentrators
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?