Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104916
Material:
polycarbonate housing, polyethersulfone membrane
Pore size:
50 kDa MWCO
Working volume:
500 μL
Sterility:
non-sterile
Packaging:
package of 25 ea
Pomoc techniczna
Pozwól nam pomócmaterial
polycarbonate housing, polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 25 ea
manufacturer/tradename
Sartorius VS0131
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
2.2 mL
working volume
500 μL
pore size
50 kDa MWCO
General description
- Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
- Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
- Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
Still not finding the right product?
Explore all of our products under Vivaspin 500 centrifugal concentrators
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?