feature
Holds 15 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 1347
technique(s)
centrifugation: suitable
compatibility
for use with 1481 1490, 1495 (buckets)
General description
- compatible with bucket/carriers 1381 (Z601543) and 1430 (Z729132)
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, Adapter for 15 mL Falcon tubes (17x120 mm) | description for ROTINA 38/38R and ROTINA 420/420R Centrifuges, Adapter for 50 mL Falcon tubes (29x115 mm) | description for ROTOFIX 32A and UNIVERSAL 320/320R, Adapter (rubber insert) for 100 mL tubes (44x100 mm) | description for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R, for 4.5-5 mL blood/urine tubes (11x92 mm) and 7 mL round bottom tubes (12x100 mm) |
| manufacturer/tradename Hettich 1347 | manufacturer/tradename Hettich 1384 | manufacturer/tradename Hettich 0761 | manufacturer/tradename Hettich 5247 |
| compatibility for use with 1481 1490, 1495 (buckets) | compatibility for use with 1481 1490, 1495 (buckets) | compatibility for use with 1481, 1490, 1495 (bucket) | compatibility for use with 5051 (buckets) |
| packaging pack of 4 ea | packaging pack of 4 ea | packaging pack of 1 ea | packaging pack of 8 ea |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
| feature Holds 15 mL tubes | feature Holds 50 mL tubes | feature Holds 100 mL tubes | feature Holds 7 mL tubes |
Still not finding the right product?
Explore all of our products under Hettich® centrifuge adapters
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?