Informacje o tej pozycji
NACRES:
NB.82
UNSPSC Code:
41103911
eCl@ss:
32110390
Compatibility:
for use with 5051 (buckets)
Manufacturer/tradename:
Hettich 5247
feature
Holds 7 mL tubes
packaging
pack of 8 ea
manufacturer/tradename
Hettich 5247
technique(s)
centrifugation: suitable
compatibility
for use with 5051 (buckets)
General description
- compatible with bucket/carriers 5051 and 5628
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Still not finding the right product?
Explore all of our products under Hettich® centrifuge adapters
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?