Przejdź do zawartości
Z380199
Test tube half racks for orbital mixer
10-13mm tubes 6 × 7 array
Synonim(y):
187480013, F18748-0013, compact rack for orbital mixers, compact tube rack, tube holder, tube rack
Informacje o tej pozycji
NACRES:
NB.73
UNSPSC Code:
41122804
Manufacturer/tradename:
Bel-Art F18748-0013
Feature:
10-13mm tubes 6 × 7 array
Packaging:
pack of 1 ea
Pomoc techniczna
Pozwól nam pomóc
feature
10-13mm tubes 6 × 7 array
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18748-0013
size
128 mm × 105 mm × 72 mm
General description
Compact size polypropylene plastic racks save space in water baths, laminar flow hoods and fume hoods.
Application
For secure orbital mixing of samples in test tubes.
Features and Benefits
  • Available in 5 grid sizes to accommodate the full range of laboratory test tubes.
  • Steam autoclavable at 121ºC (250ºF).
  • Dimensions: 12.8 x 10.5 x 7.2cmH (5.1 x 4.1 x 2.8").
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)