Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 pkg | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 354,00 zł |
Informacje o tej pozycji
354,00 zł
material
polypropylene cap, polypropylene copolymer bottle (PPCO), square translucent bottle
joint
Joints threaded neck (63)
description
Autoclavable
feature
with cap
packaging
pkg of 6
manufacturer/tradename
Nalgene® 2110-0032
bottle diam. × H
95 mm × 181 mm
capacity
1,000 mL
General description
Features and Benefits
- Made from polypropylene copolymers.
- Ideal for storing and transporting various samples.
- The square design is convenient for labeling.
- High-quality laboratory-grade materials for low leachable and extractable.
- Linerless polypropylene closures for sample protection.
- Autoclavable.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 1,000 mL | description capacity 250 mL | description size 60 mL | description capacity 175 mL |
| manufacturer/tradename Nalgene® 2110-0032 | manufacturer/tradename Nalgene® 2110-0008 | manufacturer/tradename Nalgene® 2110-0002 | manufacturer/tradename Nalgene® 2110-0006 |
| feature with cap | feature with cap | feature with cap, autoclavable | feature with cap |
| capacity 1,000 mL | capacity 250 mL | capacity - | capacity 175 mL |
| material polypropylene copolymer bottle (PPCO), polypropylene cap, square translucent bottle | material square translucent bottle, polypropylene copolymer bottle (PPCO), polypropylene cap | material polypropylene copolymer bottle (PPCO), polypropylene cap, translucent bottle, wide-mouth bottle, square bottle | material polypropylene copolymer bottle (PPCO), polypropylene cap, square translucent bottle |
| joint Joints threaded neck (63) | joint Joints threaded neck (43) | joint Joints threaded neck (28) | joint Joints threaded neck (38) |
| bottle diam. × H 95 mm × 181 mm | bottle diam. × H 61 mm × 116 mm | bottle diam. × H - | bottle diam. × H - |
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2110
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?