Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 pkg | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 124,00 zł 105,40 zł |
Informacje o tej pozycji
105,40 zł
Cena katalogowa124,00 złZaoszczędź 15%material
polypropylene cap, polypropylene copolymer bottle (PPCO), square bottle, translucent bottle, wide-mouth bottle
joint
Joints threaded neck (28)
description
Autoclavable
feature
autoclavable, with cap
packaging
pkg of 12
manufacturer/tradename
Nalgene® 2110-0002
diam. × H
36 mm × 83 mm
size
60 mL
General description
Features and Benefits
- Made from polypropylene copolymers.
- Ideal for storing and transporting various samples.
- The square design is convenient for labeling.
- High-quality laboratory-grade materials for low leachable and extractable.
- Linerless polypropylene closures for sample protection.
- Autoclavable.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description size 60 mL | description capacity 175 mL | description capacity 250 mL | description capacity 1,000 mL |
| feature with cap, autoclavable | feature with cap | feature with cap | feature with cap |
| material polypropylene copolymer bottle (PPCO), polypropylene cap, wide-mouth bottle, translucent bottle, square bottle | material polypropylene copolymer bottle (PPCO), polypropylene cap, square translucent bottle | material square translucent bottle, polypropylene copolymer bottle (PPCO), polypropylene cap | material polypropylene copolymer bottle (PPCO), square translucent bottle, polypropylene cap |
| joint Joints threaded neck (28) | joint Joints threaded neck (38) | joint Joints threaded neck (43) | joint Joints threaded neck (63) |
| packaging pkg of 12 | packaging pkg of 12 | packaging pkg of 12 | packaging pkg of 6 |
| size 60 mL | size - | size - | size - |
| diam. × H 36 mm × 83 mm | diam. × H - | diam. × H - | diam. × H - |
Still not finding the right product?
Explore all of our products under Nalgene® bottles, style 2110
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?