Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
48 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1520,00 zł |
Informacje o tej pozycji
1520,00 zł
material
high-density polyethylene cap, square bottle, transparent PETG
joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations, with cap
packaging
pack of 48 ea (2 x pack of 24)
manufacturer/tradename
Nalgene® 2019-0125
capacity
125 mL
General description
- Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
- Designed for storing buffers, cultures, and culture media for various laboratory applications.
- Features a leakproof closure for sample protection.
- Heat-shrink band around closure and neck facilitate tamper-evident seal.
- Ideal for refrigerated/frozen storage down to -40°C.
Features and Benefits
- Avoids depyrogenation, expensive washing, and autoclaving steps
- Sterile and non-pyrogenic
- Obtained in low particulate format
- Validated by extensive documentation
- Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
- Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 125 mL | description capacity 250 mL | description capacity 2000 mL | description capacity 1000 mL |
| feature graduations, with cap | feature with cap, graduations | feature graduations, with cap | feature graduations: no, with cap |
| capacity 125 mL | capacity 250 mL | capacity 2000 mL | capacity 1000 mL |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| material square bottle, high-density polyethylene cap, transparent PETG | material high-density polyethylene cap, square bottle, transparent PETG | material square bottle, transparent PETG | material high-density polyethylene cap, square bottle, transparent PETG |
| packaging pack of 48 ea (2 x pack of 24) | packaging pack of 48 ea (2 x pack of 24) | packaging pack of 12 ea (2 xpack of 6) | packaging pack of 24 ea (2 xpack of 12) |
| manufacturer/tradename Nalgene® 2019-0125 | manufacturer/tradename Nalgene® 2019-0250 | manufacturer/tradename Nalgene® 2019-2000 | manufacturer/tradename Nalgene® 342020-1000 |
Still not finding the right product?
Explore all of our products under Nalgene® media bottles, style 2019
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?