Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
12 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1650,00 zł |
Informacje o tej pozycji
1650,00 zł
material
square bottle, transparent PETG
joint
Joints threaded neck (53B)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations, with cap
packaging
pack of 12 ea (2 xpack of 6)
manufacturer/tradename
Nalgene® 2019-2000
capacity
2000 mL
General description
- Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
- Designed for storing buffers, cultures, and culture media for various laboratory applications.
- Features a leakproof closure for sample protection.
- Heat-shrink band around closure and neck facilitate tamper-evident seal.
- Ideal for refrigerated/frozen storage down to -40°C.
Features and Benefits
- Avoids depyrogenation, expensive washing, and autoclaving steps
- Sterile and non-pyrogenic
- Obtained in low particulate format
- Validated by extensive documentation
- Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
- Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 2000 mL | description capacity 1,000 mL | description capacity 500 mL | description capacity 250 mL |
| capacity 2000 mL | capacity 1,000 mL | capacity 500 mL | capacity 250 mL |
| sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated | sterility sterile; γ-irradiated |
| feature graduations, with cap | feature graduations, with cap | feature graduations, with cap | feature graduations, with cap |
| material transparent PETG , square bottle | material square bottle, high-density polyethylene cap, transparent PETG | material high-density polyethylene cap, transparent PETG , square bottle | material transparent PETG , high-density polyethylene cap, square bottle |
| packaging pack of 12 ea (2 xpack of 6) | packaging pack of 24 ea (2 xpack of 12) | packaging pack of 24 ea (2 xpack of 12) | packaging pack of 48 ea (2 x pack of 24) |
| joint Joints threaded neck (53B) | joint Joints threaded neck (38-430mm) | joint Joints threaded neck (38-430mm) | joint Joints threaded neck (38-430mm) |
Still not finding the right product?
Explore all of our products under Nalgene® media bottles, style 2019
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?