Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
4 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 518,00 zł 440,30 zł |
Informacje o tej pozycji
440,30 zł
Cena katalogowa518,00 złZaoszczędź 15%material
colorless polypropylene cap, flat bottom, polymethylpentene , wide-neck
joint
Joints neck (43)
sterility
non-sterile
feature
closure type Screw cap, with cap
packaging
pack of 4
manufacturer/tradename
Nalgene 4109-0500
capacity
500 mL
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 500 mL | description capacity 1,000 mL | description capacity 250 mL | description capacity 125 mL |
| capacity 500 mL | capacity 1,000 mL | capacity 250 mL | capacity 125 mL |
| manufacturer/tradename Nalgene 4109-0500 | manufacturer/tradename Nalgene 4109-1000 | manufacturer/tradename Nalgene 4109-0250 | manufacturer/tradename Nalgene 4109-0125 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material polymethylpentene , flat bottom, wide-neck , colorless polypropylene cap | material polymethylpentene , colorless polypropylene cap, wide-neck , flat bottom | material narrow-mouth , polymethylpentene , flat bottom, colorless polypropylene cap | material colorless polypropylene cap, flat bottom, polymethylpentene , wide-neck |
| feature with cap, closure type Screw cap | feature closure type Screw cap, with cap | feature closure type Screw cap, with cap | feature closure type Screw cap, with cap |
| joint Joints neck (43) | joint Joints neck (53) | joint Joints neck (38) | joint Joints neck (33) |
Still not finding the right product?
Explore all of our products under Nalgene® Polymethylpentene Erlenmeyer flask
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?