Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
4 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 322,00 zł 273,70 zł |
Informacje o tej pozycji
273,70 zł
Cena katalogowa322,00 złZaoszczędź 15%material
colorless polypropylene cap, flat bottom, narrow-mouth , polymethylpentene
joint
Joints neck (38)
sterility
non-sterile
feature
closure type Screw cap, with cap
packaging
pack of 4
manufacturer/tradename
Nalgene 4109-0250
capacity
250 mL
General description
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description capacity 250 mL | description capacity 125 mL | description capacity 250 mL | description capacity 125 mL |
| manufacturer/tradename Nalgene 4109-0250 | manufacturer/tradename Nalgene 4109-0125 | manufacturer/tradename Nalgene 4102-0250 | manufacturer/tradename Nalgene 4103-0125 |
| capacity 250 mL | capacity 125 mL | capacity 250 mL | capacity 125 mL |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| material narrow-mouth , flat bottom, polymethylpentene , colorless polypropylene cap | material wide-neck , colorless polypropylene cap, polymethylpentene , flat bottom | material polypropylene copolymer bottle (PPCO), flat bottom | material polycarbonate , flat bottom, wide-neck |
| feature with cap, closure type Screw cap | feature with cap, closure type Screw cap | feature cap: no, without cap | feature without cap, closure type Stoppers |
| joint Joints neck (38) | joint Joints neck (33) | joint Joints stopper (6) | joint joint stopper5 |
Still not finding the right product?
Explore all of our products under Nalgene® Polymethylpentene Erlenmeyer flask
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?