Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
2 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 283,00 zł |
Informacje o tej pozycji
283,00 zł
material
borosilicate glass 3.3 flask
Quality Segment
joint
joint ST/NS neck12/21
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37448
volume measuring range
50 mL, accuracy: 0.06 mL
capacity
50 mL
General description
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description volume 50 mL, accuracy: 0.06 mL | description volume 50 mL, accuracy: 0.06 mL | description volume 5 mL, accuracy: 0.08 mL | description volume 10 mL, accuracy: 0.04 mL |
| capacity 50 mL | capacity 50 mL | capacity 5 mL | capacity 10 mL |
| feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In)., closure type Glass stopper | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). | feature class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In). |
| manufacturer/tradename BRAND 37448 | manufacturer/tradename BRAND | manufacturer/tradename BRAND 37438 | manufacturer/tradename BRAND 37443 |
| joint joint ST/NS neck12/21 | joint joint ST/NS neck14/23 | joint joint ST/NS neck10/19 | joint joint ST/NS neck10/19 |
| material borosilicate glass 3.3 flask | material borosilicate glass , wide-neck | material borosilicate glass 3.3 flask, wide-neck | material borosilicate glass 3.3 flask, wide-neck |
| packaging pack of 2 ea | packaging pack of 2 | packaging pack of 2 ea | packaging pack of 2 ea |
Still not finding the right product?
Explore all of our products under BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?