Przejdź do zawartości
Z327018
BRAND® BLAUBRAND® volumetric flask, glass stopper, amber glass
volume 10 mL, accuracy: 0.04 mL
Synonim(y):
amber flask, analytical flask, calibration flask with stopper, measuring flask, volumetric flask
Informacje o tej pozycji
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Capacity:
10 mL
Material:
borosilicate glass 3.3 flask, wide-neck
Packaging:
pack of 2 ea
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
material
borosilicate glass 3.3 flask, wide-neck
joint
joint ST/NS neck10/19
feature
class A DE-M, DIN EN ISO 1042. Calibrated to contain (TC, In).
packaging
pack of 2 ea
manufacturer/tradename
BRAND 37443
volume measuring range
10 mL, accuracy: 0.04 mL
capacity
10 mL
General description
Volumetric flasks according to DIN EN ISO 1042, Class A, conformity certified. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a interchangeable amber glass stopper. Marks and inscriptions in high contrast white enamel.
All BLAUBRAND® volumetric flasks are supplied with a batch number and an accompanying batch certificate in the original packaging.
Other Notes
BR37443 (India)
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)