Przejdź do
Z290408
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with glass support, NS 40/35 joints, 1 L flask (included)
Synonim(y):
Filtration Assembly, Vacuum Filtration System
, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
2850,00 zł
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
2850,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
glass housing, glass membrane
joint
joint ST/NS40/35
feature
graduation
packaging
pkg of 1 ea
bottle capacity
1 L
funnel capacity
300 mL
General description
Same as Z290432 but with NS 40/35 joint (male on flask, female on funnel support) instead of rubber stopper.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z290467Z290424Z290475
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
feature
graduation
feature
cap plug seal, graduation
feature
graduation
feature
cap plug seal, graduation
joint
joint ST/NS40/35
joint
-
joint
joint ST/NS40/35
joint
-
material
glass housing, glass membrane
material
glass housing, glass membrane
material
glass housing, glass membrane
material
glass housing, glass membrane
bottle capacity
1 L
bottle capacity
125 mL
bottle capacity
4 L
bottle capacity
-
funnel capacity
300 mL
funnel capacity
15 mL
funnel capacity
1 L
funnel capacity
15 mL
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)