Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1530,00 zł |
Informacje o tej pozycji
1530,00 zł
sterility
sterile
feature
filter 330 μm
packaging
pkg of 100 ea
manufacturer/tradename
Whirl-Pak® B01525WA
W × L
38 cm × 38 cm , 15" x 15"
application(s)
sample preparation
compatibility
for use with Whirl-pak® homogenizer
General description
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate; bags should not be used at temperatures above 180° F, 82° C. Bags can be frozen to any temperature, even liquid nitrogen - careful handling required after freezing. All bags are sterilized after manufacturing.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description W × L 38 cm × 38 cm , 15" x 15" | description W × L 25.4 cm × 38 cm , 10" x 15" | description 2041 mL capacity(69 oz) | description 710 mL capacity(24 oz) |
| feature filter 330 μm | feature filter 330 μm | feature filter 330 μm | feature filter 330 μm |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Whirl-Pak® B01525WA | manufacturer/tradename Whirl-Pak® B01488WA | manufacturer/tradename Whirl-Pak® B01416WA | manufacturer/tradename Whirl-Pak® B01348WA |
| application(s) sample preparation | application(s) sample preparation | application(s) - | application(s) sample preparation |
| W × L 38 cm × 38 cm , 15" x 15" | W × L 25.4 cm × 38 cm , 10" x 15" | W × L - | W × L 6 in. × 9 in. , 15 x 23 cm |
| packaging pkg of 100 ea | packaging pkg of 100 ea | packaging pkg of 250 ea | packaging pkg of 250 ea |
Still not finding the right product?
Explore all of our products under Whirl-Pak® Homogenizer Blender Filter Bag
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?