Merck
R5776
20 well tube rack
blue
Informacje o tej pozycji
NACRES:
NB.35
UNSPSC Code:
41122804
Manufacturer/tradename:
Heathrow Scientific HS4284A, Heathrow Scientific
Feature:
Holds 20 x 1-2 mL tubes, Holds 20 x 10 mm tubes
Material:
polypropylene
Pomoc techniczna
Pozwól nam pomóc
material
polypropylene
description
Autoclavable
feature
Holds 20 x 1-2 mL tubes, Holds 20 x 10 mm tubes
manufacturer/tradename
Heathrow Scientific HS4284A, Heathrow Scientific
size
211x90x50 mm
color
blue
General description
These racks for 1.5-2.0 mL tubes slide together on both sides to permit the user to attach several for larger experiments. They feature an innovative "hold-it" finger slot on the front, and internal ribbing for stability.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)