Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 220,00 zł |
Informacje o tej pozycji
220,00 zł
material
blue , high-density polyethylene coating
feature
to hold 80 x 20-25 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120768
L × W × H
11.4 in. (29 cm) × 9.1 in. (23 cm) × 3.1 in. (8 cm)
General description
Features and Benefits
- Chemically-resistant HDPE Coated Wire Racks
- Racks can be used between temperatures of -40°C and 121°C
- Holds all brands and types of conical, round or flat bottom, glass or plastic, tubes and/or bottles
- Square wells and three deck levels help keep tubes secure
- Autoclavable
- Mesh Bottom Support
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description to hold, 80 x 20-25 mm tubes, blue | description to hold, 108 x 13-16 mm tubes, blue | description to hold, 48 x 25-30 mm tubes, blue | description to hold, 40 x 16-20 mm tubes, blue |
| manufacturer/tradename Heathrow Scientific 120768 | manufacturer/tradename Heathrow Scientific 120763 | manufacturer/tradename Heathrow Scientific 120770 | manufacturer/tradename Heathrow Scientific 120764 |
| feature to hold 80 x 20-25 mm tubes | feature to hold 108 x 13-16 mm tubes | feature to hold 48 x 25-30 mm tubes | feature to hold 40 x 16-20 mm tubes |
| material blue , high-density polyethylene coating | material blue , high-density polyethylene coating | material blue , high-density polyethylene coating | material blue , high-density polyethylene coating |
| L × W × H 11.4 in. (29 cm) × 9.1 in. (23 cm) × 3.1 in. (8 cm) | L × W × H 9.3 in. (23.7 cm) × 5 in. (12.7 cm) × 2.6 in. (6.5 cm) | L × W × H 10.9 in. (27.7 cm) × 8.3 in. (20.9 cm) × 3.9 in. (9.8 cm) | L × W × H 9.5 in. (24 cm) × 3.9 in. (9 cm) × 3.1 in. (8 cm) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Coated Wire Tube Rack
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?