Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 164,00 zł |
Informacje o tej pozycji
164,00 zł
material
acrylonitrile-butadiene-styrene (ABS) , purple (dispenser)
feature
to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120668
L × W × H
123 mm × 172 mm × 99 mm
General description
Features and Benefits
- Accommodates two 50 mm (2-inch), or one 100 mm (4-inch) Parafilm® M rolls
- Easy application, simply place the dispenser on top of the Parafilm® M box and pull the sealing film through, there is no need to remove rolls from the packaging
- Built in safety blade slides and cuts with no danger to fingertips and eliminates the need for scissors
- Supplied with two safety blades, additional replacement blades are available for purchase
- Made from ABS plastic making the unit easy to clean
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| feature to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll | feature to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll | feature to hold 2 x 50 mm or 1 x 100 mm Parafilm M Sealing Film roll | feature holds 2 x 50 mm or 1 x 100 mm ParafilmM Sealing Film roll |
| manufacturer/tradename Heathrow Scientific 120668 | manufacturer/tradename Heathrow Scientific 120667 | manufacturer/tradename Heathrow Scientific 120758 | manufacturer/tradename Heathrow Scientific HS234524 |
| material acrylonitrile-butadiene-styrene (ABS) , purple (dispenser) | material acrylonitrile-butadiene-styrene (ABS) , blue (dispenser) | material acrylonitrile-butadiene-styrene (ABS) , natural (dispenser) | material acrylic , clear (dispenser) |
| L × W × H 123 mm × 172 mm × 99 mm | L × W × H 123 mm × 172 mm × 99 mm | L × W × H 123 mm × 172 mm × 99 mm | L × W × H 119 mm × 168 mm × 175 mm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
Still not finding the right product?
Explore all of our products under Box Top Dispenser for Parafilm
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?