Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
500 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 214,00 zł |
Informacje o tej pozycji
214,00 zł
material
square blue polystyrene
size
small
feature
Anti-static
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS120222
L × W × H
(46 mm) × 46 mm × 8 mm
General description
Features and Benefits
- biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- temperature range -10 to 70C
- disposable
- shallow with a wide, flat bottom to resist tipping
- rounded corners simplify transfer
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Small, square blue polystyrene, Anti-static, pk of 500 ea | description Large, square blue polystyrene, Anti-static, pk of 500 ea | description Medium, square blue polystyrene, Anti-static, pk of 500 ea | description Small, square blue polystyrene, pk of 500 ea |
| manufacturer/tradename Heathrow Scientific HS120222 | manufacturer/tradename Heathrow Scientific HS120224 | manufacturer/tradename Heathrow Scientific HS120223 | manufacturer/tradename Heathrow Scientific HS1421A |
| material square blue polystyrene | material square blue polystyrene | material square blue polystyrene | material square blue polystyrene |
| packaging pk of 500 ea | packaging pk of 500 ea | packaging pk of 500 ea | packaging pk of 500 ea |
| size small | size large | size medium | size small |
| feature Anti-static | feature Anti-static | feature Anti-static | feature - |
| L × W × H (46 mm) × 46 mm × 8 mm | L × W × H (140 mm) × 140 mm × 22 mm | L × W × H (85 mm) × 84 mm × 24 mm | L × W × H - |
Still not finding the right product?
Explore all of our products under Weigh Boat
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?